3-Oxa-2-thia-1-azaspiro[4.4]nonane 2,2-dioxide, N-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

OR52997-1G OR52997-250MG OR52997-5G
APOSOR52997-1GEA 145 EUR
APOSOR52997-1G APOSOR52997-250MG APOSOR52997-5G
3-Oxa-2-thia-1-azaspiro[4.4]nonane 2,2-dioxide, N-BOC protected 95%
3-Oxa-2-thia-1-azaspiro[4.4]nonane 2,2-dioxide, N-BOC protected
Formel: C₁₁H₁₉NO₅S
Lagertemperatur: Kühlschrank
MDL: MFCD31743878
CAS-Nummer: 2306264-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR