3-Bromo-5-ethynylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromopyridin-3-yl)acetylene

OR53006-250MG
APOSOR53006-250MGEA 161 EUR
APOSOR53006-250MG
3-Bromo-5-ethynylpyridine
3-Bromo-5-ethynylpyridine
Formel: C₇H₄BrN
Molecular Weight: 182,017 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10698137
CAS-Nummer: 866683-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR