5-Ethynylpyridine-2-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Ethynylpicolinonitrile, 2-Cyano-5-ethynylpyridine

OR53008-250MG
APOSOR53008-250MGEA 161 EUR
APOSOR53008-250MG
5-Ethynylpyridine-2-carbonitrile
5-Ethynylpyridine-2-carbonitrile
Formel: C₈H₄N₂
Molecular Weight: 128,131 g/mol
Schmelzpunkt: 126…127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD18205188
CAS-Nummer: 1211584-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR