6-Ethynylimidazo[1,2-a]pyrazine
Lieferant: Apollo Scientific
|
Achtung
|
OR53013-1G
OR53013-250MG
APOSOR53013-1GEA
0
EUR
APOSOR53013-1G
APOSOR53013-250MG
6-Ethynylimidazo[1,2-a]pyrazine
6-Ethynylimidazo[1,2-a]pyrazine
Formel:
C₈H₅N₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD21647858 CAS-Nummer: 1372096-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...