2-Ethynyl-5-methoxypyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Ethynyl-5-methoxy-1,4-diazine, (5-Methoxypyrazin-2-yl)acetylene

OR53017-250MG
APOSOR53017-250MGEA 143 EUR
APOSOR53017-250MG
2-Ethynyl-5-methoxypyrazine
2-Ethynyl-5-methoxypyrazine
Formel: C₇H₆N₂O
Molecular Weight: 134,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22199320
CAS-Nummer: 1374115-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR