N,N-dimethylpyrrolidine-3-sulphonamide hydrochloride

Lieferant: Apollo Scientific
Achtung

OR53025-250MG
APOSOR53025-250MGEA 546 EUR
APOSOR53025-250MG
N,N-dimethylpyrrolidine-3-sulphonamide hydrochloride
N,N-dimethylpyrrolidine-3-sulphonamide hydrochloride
Formel: C₆H₁₄N₂O₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27756580
CAS-Nummer: 1825309-05-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR