8-Bromo-1,6-naphthyridin-5-amine 97%
Lieferant: Apollo Scientific
OR53039-250MG
APOSOR53039-250MGEA
92.8
EUR
APOSOR53039-250MG
8-Bromo-1,6-naphthyridin-5-amine 97%
8-Bromo-1,6-naphthyridin-5-amine
Formel:
C₈H₆BrN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22396116 CAS-Nummer: 1820686-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...