2,5-Dichlor-3-nitrobenzoesäure 95%

Lieferant: Apollo Scientific
Achtung

OR53044-5G
APOSOR53044-5GEA 166 EUR
APOSOR53044-5G
2,5-Dichlor-3-nitrobenzoesäure 95%
2,5-Dichloro-3-nitrobenzoic acid
Formel: C₇H₃Cl₂NO₄
Molecular Weight: 236.01 g/mol
Schmelzpunkt: 216…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014691
CAS-Nummer: 88-86-8
EINECS: 201-862-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR