4-Hydroxy-3-methoxy-5-methylbenzonitrile 95+%

Lieferant: Apollo Scientific

OR53049-250MG
APOSOR53049-250MGEA 48 EUR
APOSOR53049-250MG
4-Hydroxy-3-methoxy-5-methylbenzonitrile 95+%
4-Hydroxy-3-methoxy-5-methylbenzonitrile
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18432784
CAS-Nummer: 173900-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR