4-(Isocyanatomethyl)tetrahydropyran
Lieferant: Apollo Scientific
|
Gefahr
|
OR53052-500MG
APOSOR53052-500MGEA
575
EUR
APOSOR53052-500MG
4-(Isocyanatomethyl)tetrahydropyran
4-(Isocyanatomethyl)tetrahydropyran
- May form small amount of sediment.
Formel:
C₇H₁₁NO₂ Molecular Weight: 141,17 g/mol Siedepunkt: 110…115 °C/20mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD09817556 CAS-Nummer: 934570-48-6 UN: 2478 ADR: 3+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...