5-Ethynylimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (Imidazo[1,2-a]pyridin-5-yl)acetylene

OR53053-250MG
APOSOR53053-250MGEA 285 EUR
APOSOR53053-250MG
5-Ethynylimidazo[1,2-a]pyridine
5-Ethynylimidazo[1,2-a]pyridine
Formel: C₉H₆N₂
Molecular Weight: 142,157 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22199325
CAS-Nummer: 1374115-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR