4-Hydroxy-2-nitrobenzonitrile 95%

Lieferant: Apollo Scientific

OR53078-250MG
APOSOR53078-250MGEA 214 EUR
APOSOR53078-250MG
4-Hydroxy-2-nitrobenzonitrile 95%
4-Hydroxy-2-nitrobenzonitrile
Formel: C₇H₄N₂O₃
Molecular Weight: 164,118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527342
CAS-Nummer: 1093203-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR