Ethyl 3-(4-methoxyphenyl)propanoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl-p-methoxyhydrocinnamate

OR53089-1G
APOSOR53089-1GEA 143 EUR
APOSOR53089-1G
Ethyl 3-(4-methoxyphenyl)propanoate 98%
Ethyl 3-(4-methoxyphenyl)propanoate
Formel: C₁₂H₁₆O₃
Molecular Weight: 208,254 g/mol
Schmelzpunkt: 52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258632
CAS-Nummer: 22767-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR