2,3-Dihydro[1,4]dioxino[2,3-b]pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-1,4-dioxino[2,3-b]pyridine

OR53142-1G
APOSOR53142-1GEA 75.4 EUR
APOSOR53142-1G
2,3-Dihydro[1,4]dioxino[2,3-b]pyridine 97%
2,3-Dihydro[1,4]dioxino[2,3-b]pyridine
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09743547
CAS-Nummer: 129421-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR