Methyl 5-phenylnicotinate

Lieferant: Apollo Scientific
Achtung

OR53146-1G
APOSOR53146-1GEA 230 EUR
APOSOR53146-1G
Methyl 5-phenylnicotinate
Methyl 5-phenylnicotinate
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,232 g/mol
Siedepunkt: 356 °C (760 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD06802742
CAS-Nummer: 10177-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR