4-Brom-4'-nitrobiphenyl 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-4'-nitro-1,1'-biphenyl

OR53160-1G
APOSOR53160-1GEA 78.3 EUR
APOSOR53160-1G
4-Brom-4'-nitrobiphenyl 95+%
4-Brom-4'-nitrobiphenyl
Formel: C₁₂H₈BrNO₂
Molecular Weight: 278,1014 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024697
CAS-Nummer: 6242-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR