trans-2-Phenylcyclopropylamine hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR53161-1G
APOSOR53161-1GEA 112 EUR
APOSOR53161-1G
trans-2-Phenylcyclopropylamine hydrochloride
trans-2-Phenylcyclopropylamine hydrochloride
Formel: C₉H₁₂ClN
Lagertemperatur: Kühlschrank
MDL: MFCD00063602
CAS-Nummer: 1986-47-6
UN: 3249
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR