Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-amino-1,3,4-oxadiazole-2-carboxylate

OR53168-1G OR53168-5G
APOSOR53168-1GEA 21.9 EUR
APOSOR53168-1G APOSOR53168-5G
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate 98%
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate
Formel: C₅H₇N₃O₃
Molecular Weight: 157.13 g/mol
Schmelzpunkt: 200…205 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03425200
CAS-Nummer: 4970-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR