Ethyl acetoacetate propylene glycol ketal 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl Acetoacetate Propylene glycol Ketal, 2-(2,4-Dimethyl-1,3-dioxolan-2-yl)acetic acid ethyl ester, Ethyl 2-(2,4-Dimethyl-1,3-dioxolan-2-yl)acetate (mixture of diastereoisomers)

OR53171-100G
APOSOR53171-100GEA 77 EUR
APOSOR53171-100G
Ethyl acetoacetate propylene glycol ketal 98+%
Ethyl acetoacetate propylene glycol ketal
Formel: C₉H₁₆O₄
Molecular Weight: 188,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151819
CAS-Nummer: 6290-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR