1,5-Naphthyridin-3-ol 95%

Lieferant: Apollo Scientific
Achtung

OR53176-100MG OR53176-250MG
APOSOR53176-100MGEA 340 EUR
APOSOR53176-100MG APOSOR53176-250MG
1,5-Naphthyridin-3-ol 95%
1,5-Naphthyridin-3-ol
Formel: C₈H₆N₂O
Molecular Weight: 146.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656382
CAS-Nummer: 14756-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR