3-Azabicyclo[3.3.0]octane hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Octahydrocyclopenta[c]pyrrole hydrochloride

OR53182-100G
APOSOR53182-100GEA 74.1 EUR
APOSOR53182-100G
3-Azabicyclo[3.3.0]octane hydrochloride
3-Azabicyclo[3.3.0]octane hydrochloride
Formel: C7H14ClN
Molecular Weight: 147.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411019
CAS-Nummer: 112626-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR