3-Biphenyl isothiocyanate 97%
Lieferant: Apollo Scientific
Synonyme:
3-Isothiocyanato-1,1'-biphenyl
OR53213-250MG
APOSOR53213-250MGEA
183
EUR
APOSOR53213-250MG
3-Biphenyl isothiocyanate 97%
3-Biphenyl isothiocyanate
Formel:
C₁₃H₉NS Molecular Weight: 211,29 g/mol Siedepunkt: 150 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025658 CAS-Nummer: 1510-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...