3-Amino-5-(tert-butyl)thiophene-2-carboxamide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-(tert-butyl)-2-carbamoylthiophene, 3-Amino-2-(aminocarbonyl)-5-(tert-butyl)thiophene

OR5325-100MG
APOSOR5325-100MGEA 27.7 EUR
APOSOR5325-100MG
3-Amino-5-(tert-butyl)thiophene-2-carboxamide 98%
3-Amino-5-(tert-butyl)thiophene-2-carboxamide
Formel: C₉H₁₄N₂OS
Molecular Weight: 198,285 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00068165
CAS-Nummer: 175137-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR