2,6-Dichlorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,6-Dichlorphenyl)metaborsäure, 2,6-Dichlorbenzolboronsäure, Dihydroxy(2,6-dichlorphenyl)boran

OR5330-25G OR5330-5G OR5330-1G OR5330-10G
APOSOR5330-25GEA 46.8 EUR
APOSOR5330-25G APOSOR5330-5G APOSOR5330-1G APOSOR5330-10G
2,6-Dichlorphenylboronsäure 98%
2,6-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190.82 g/mol
Schmelzpunkt: 151…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064869
CAS-Nummer: 73852-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR