3-(2-Naphthyl)acrylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR5352-25G OR5352-1G OR5352-5G
APOSOR5352-25GEA 403 EUR
APOSOR5352-25G APOSOR5352-1G APOSOR5352-5G
3-(2-Naphthyl)acrylic acid 98%
3-(2-Naphthyl)acrylic acid
Formel: C₁₃H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00209666
CAS-Nummer: 51557-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR