4-(Methoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid

OR5354-100G OR5354-25G OR5354-5G
APOSOR5354-100GEA 108 EUR
APOSOR5354-100G APOSOR5354-25G APOSOR5354-5G
4-(Methoxycarbonyl)phenylboronsäure 98%
4-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.97 g/mol
Schmelzpunkt: 197…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632203
CAS-Nummer: 99768-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR