1,1-Cyclobutandicarbonsäure 98%
Lieferant: Apollo Scientific
Synonyme:
Cyclobutane-1,1-dicarboxylic acid
OR5361-100G
APOSOR5361-100GEA
71.2
EUR
APOSOR5361-100G
1,1-Cyclobutandicarbonsäure 98%
1,1-Cyclobutandicarbonsäure
Formel:
C₆H₈O₄ Molecular Weight: 144,13 g/mol Schmelzpunkt: 158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001325 CAS-Nummer: 5445-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...