5-Chloro-3-methylbenzo[b]thiophene-2-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR5377-1G
APOSOR5377-1GEA
21.9
EUR
APOSOR5377-1G
5-Chloro-3-methylbenzo[b]thiophene-2-sulphonyl chloride
5-Chloro-3-methylbenzo[b]thiophene-2-sulphonyl chloride
Formel:
C₉H₆Cl₂O₂S₂ Schmelzpunkt: 153 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00173797 CAS-Nummer: 166964-33-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...