(S)-Methyl N-tert-butoxycarbonyl-3-(4-biphenylyl)-2-aminopropionate 98%

Lieferant: Apollo Scientific

Synonyme: (S)-Methyl 3-([1,1'-biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoate

OR5402-5G
APOSOR5402-5GEA 2560 EUR
APOSOR5402-5G
(S)-Methyl N-tert-butoxycarbonyl-3-(4-biphenylyl)-2-aminopropionate 98%
(S)-Methyl N-tert-butoxycarbonyl-3-(4-biphenylyl)-2-aminopropionate
Formel: C₂₁H₂₅NO₄
Molecular Weight: 355,427 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075194
CAS-Nummer: 137255-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR