1-Boc-4-hydroxypiperidin 98+%
Lieferant: Apollo Scientific
|
Achtung
|
OR5404-100G
OR5404-25G
APOSOR5404-100GEA
0
EUR
APOSOR5404-100G
APOSOR5404-25G
1-Boc-4-hydroxypiperidin 98+%
1-Boc-4-hydroxypiperidin
Formel:
C₁₀H₁₉NO₃ Schmelzpunkt: 61…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075174 CAS-Nummer: 109384-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...