1-Boc-piperidin-4-carbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

OR5410-25G OR5410-100G
APOSOR5410-25GEA 23.2 EUR
APOSOR5410-25G APOSOR5410-100G
1-Boc-piperidin-4-carbonsäure 98%
1-Boc-piperidin-4-carbonsäure
Formel: C₁₁H₁₉NO₄
Schmelzpunkt: 148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00076999
CAS-Nummer: 84358-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR