2-Amino-5-methoxypyrimidine

Lieferant: Apollo Scientific
Achtung

OR54119-1G
APOSOR54119-1GEA 85.7 EUR
APOSOR54119-1G
2-Amino-5-methoxypyrimidine
2-Amino-5-methoxypyrimidine
Formel: C₅H₇N₃O
Molecular Weight: 125,131 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00187864
CAS-Nummer: 13418-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR