Methyl-4-chlor-3-methylbenzoat 

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-3-methylbenzoic Acid Methyl Ester, Methyl 4-chloro-3-methylbenzoate

OR5430-1G
APOSOR5430-1GEA 21.9 EUR
APOSOR5430-1G
Methyl-4-chlor-3-methylbenzoat 
Methyl-4-chlor-3-methylbenzoat 
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Siedepunkt: 78 °C/0,06mmHg
Dichte: 1,21 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151838
CAS-Nummer: 91367-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR