Methyl-4-(methylamino)-3-nitrobenzenecarboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 4-(methylamino)-3-nitrobenzoate

OR54327-5G
APOSOR54327-5GEA 102 EUR
APOSOR54327-5G
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate
Formel: C₉H₁₀N₂O₄
Molecular Weight: 210.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09862450
CAS-Nummer: 36242-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR