2-(2-Ethoxy-2-oxoethyl)nicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR54329-1G
OR54329-250MG
APOSOR54329-1GEA
132
EUR
APOSOR54329-1G
APOSOR54329-250MG
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
Formel:
C₁₀H₁₁NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00663817 CAS-Nummer: 35969-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...