2-(2-Ethoxy-2-oxoethyl)nicotinic acid

Lieferant: Apollo Scientific
Achtung

OR54329-1G OR54329-250MG
APOSOR54329-1GEA 132 EUR
APOSOR54329-1G APOSOR54329-250MG
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
2-(2-Ethoxy-2-oxoethyl)nicotinic acid
Formel: C₁₀H₁₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00663817
CAS-Nummer: 35969-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR