Methyl-5-hydroxy-4-methoxy-2-nitrobenzoate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-hydroxy-4-methoxy-2-nitrobenzoate

OR54343-1G
APOSOR54343-1GEA 121 EUR
APOSOR54343-1G
Methyl-5-hydroxy-4-methoxy-2-nitrobenzoate 95+%
Methyl-5-hydroxy-4-methoxy-2-nitrobenzoate
Formel: C₉H₉NO₆
Molecular Weight: 227,18 g/mol
Schmelzpunkt: 146…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17168870
CAS-Nummer: 215659-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR