4-Bromo-1-benzothiophene-2-carbaldehyde ≥98%
Lieferant: Apollo Scientific
OR54346-250MG
APOSOR54346-250MGEA
0
EUR
APOSOR54346-250MG
4-Bromo-1-benzothiophene-2-carbaldehyde ≥98%
4-Bromo-1-benzothiophene-2-carbaldehyde
Formel:
C₉H₅BrOS Schmelzpunkt: 98…99 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD19381762 CAS-Nummer: 19075-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...