(4-Bromo-1-benzothiophen-2-yl)methanol

Lieferant: Apollo Scientific

OR54348-1G
APOSOR54348-1GEA 434 EUR
APOSOR54348-1G
(4-Bromo-1-benzothiophen-2-yl)methanol
(4-Bromo-1-benzothiophen-2-yl)methanol
Formel: C₉H₇BrOS
Schmelzpunkt: 74…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12032276
CAS-Nummer: 93103-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR