3-(Hydroxymethyl)benzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
m-Phenylendimethanol, m-Bis(hydroxymethyl)benzol, 1,3-Benzoldimethanol, m-Xylol-α,α'-diol, 1,3-Bis(hydroxymethyl)benzol
OR54383-25G
OR54383-5G
OR54383-100G
APOSOR54383-25GEA
84.1
EUR
APOSOR54383-25G
APOSOR54383-5G
APOSOR54383-100G
3-(Hydroxymethyl)benzylalkohol
3-(Hydroxymethyl)benzylalkohol
Formel:
C₈H₁₀O₂ Siedepunkt: 154…159 °C/13mmHg Schmelzpunkt: 56…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004649 CAS-Nummer: 626-18-6 EINECS: 210-934-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...