4-tert-Butylbenzhydrazid 98%

Lieferant: Apollo Scientific

OR5439-100G
APOSOR5439-100GEA 62.5 EUR
APOSOR5439-100G
4-tert-Butylbenzhydrazid 98%
4-tert-Butylbenzhydrazid
Formel: C₁₁H₁₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00014763
CAS-Nummer: 43100-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR