2-Aminoisophthalic acid 98%

Lieferant: Apollo Scientific
Achtung

OR5441-5G OR5441-1G OR5441-25G
APOSOR5441-5GEA 24.8 EUR
APOSOR5441-5G APOSOR5441-1G APOSOR5441-25G
2-Aminoisophthalic acid 98%
2-Aminoisophthalic acid
Formel: C₈H₇NO₄
Schmelzpunkt: 300…304 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00817808
CAS-Nummer: 39622-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR