Pargyline hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR54425-1G
APOSOR54425-1GEA 61.3 EUR
APOSOR54425-1G
Pargyline hydrochloride
Pargyline hydrochloride
Formel: C₁₁H₁₄ClN
Schmelzpunkt: 160…163 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012492
CAS-Nummer: 306-07-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR