6-Bromchinazolin

Lieferant: Apollo Scientific
Achtung

OR54428-5G
APOSOR54428-5GEA 111 EUR
APOSOR54428-5G
6-Bromchinazolin
6-Bromoquinazoline
Formel: C₈H₅BrN₂
Molecular Weight: 209.0400 g/mol
Schmelzpunkt: 304.2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08236695
CAS-Nummer: 89892-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR