4-(Methylthio)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR5443-25G
OR5443-500MG
OR5443-5G
APOSOR5443-25GEA
0
EUR
APOSOR5443-25G
APOSOR5443-500MG
APOSOR5443-5G
4-(Methylthio)phenylboronsäure
4-(Methylthio)phenylboronsäure
Formel:
C₇H₉BO₂S Schmelzpunkt: 210…214 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093410 CAS-Nummer: 98546-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...