2-Methyl-N-phenylaniline 98%

Lieferant: Apollo Scientific

OR54435-1G
APOSOR54435-1GEA 63.8 EUR
APOSOR54435-1G
2-Methyl-N-phenylaniline 98%
2-Methyl-N-phenylaniline
Formel: C₁₃H₁₃N
Molecular Weight: 183,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318642
CAS-Nummer: 1205-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR