3-Thiophenboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Thienylboronsäure, Thiophen-3-boronsäure

OR5445-5G
APOSOR5445-5GEA 29 EUR
APOSOR5445-5G
3-Thiophenboronsäure
3-Thiopheneboronic acid
Formel: C₄H₅BO₂S
Molecular Weight: 127,96 g/mol
Schmelzpunkt: 164…169 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151851
CAS-Nummer: 6165-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR