6-Chloruracil 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chlor-2,4-dihydroxypyrimidin, 4-Chlor-2,6-dihydroxypyrimidin, 4-Chloruracil, 6-Chlor-2,4(1H,3H)-pyrimidindion
OR5447-25G
APOSOR5447-25GEA
21.9
EUR
APOSOR5447-25G
6-Chloruracil 98%
6-Chloruracil
Formel:
C₄H₃ClN₂O₂ Molecular Weight: 146.53 g/mol Schmelzpunkt: 295 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014595 CAS-Nummer: 4270-27-3 EINECS: 224-258-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...