6-Chloro-5-methylpyridazin-3-amine

Lieferant: Apollo Scientific
Achtung

OR54479-5G
APOSOR54479-5GEA 94.4 EUR
APOSOR54479-5G
6-Chloro-5-methylpyridazin-3-amine
6-Chloro-5-methylpyridazin-3-amine
Formel: C₅H₆ClN₃
Lagertemperatur: Kühlschrank
MDL: MFCD19203420
CAS-Nummer: 66346-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR