N-Carbobenzoxy-1,3-diaminopropane hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, N-Z-1,3-Propanediamine hydrochloride, N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, N-Cbz-1,3-diaminopropane Hydrochloride, Benzyl N-(3-Aminopropyl)carbamate Hydrochloride, N-(3-Aminopropyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl (3-aminopropyl)carbamate hydrochloride

OR54485-5G
APOSOR54485-5GEA 98.6 EUR
APOSOR54485-5G
N-Carbobenzoxy-1,3-diaminopropane hydrochloride
N-Carbobenzoxy-1,3-diaminopropane hydrochloride
Formel: C₁₁H₁₆N₂O₂
Molecular Weight: 244.72 g/mol
Schmelzpunkt: 185…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00270153
CAS-Nummer: 17400-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR